missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butanol 95.0+%, TCI America™

Catalog No. D13335ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13335ML 5 mL
D133325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13335ML Supplier TCI America Supplier No. D13335ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 624-95-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00002928
InChI Key DUXCSEISVMREAX-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096
PubChem CID 12233
IUPAC Name 3,3-dimethylbutan-1-ol
SMILES CC(C)(C)CCO

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 mL
UN Number 2282
Formula Weight 102.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-butanol
EINECSNumber (2)-0217
TSCA Yes