missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethoxypropanenitrile, 90%

Catalog No. AC446261000
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446261000 100 mL
AC446260250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446261000 Supplier Thermo Scientific Chemicals Supplier No. 446261000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57597-62-3 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57597-62-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00044797
InChI Key JYHSJQNYYLGMEI-UHFFFAOYSA-N
PubChem CID 93731
IUPAC Name 3,3-dimethoxypropanenitrile
SMILES COC(CC#N)OC

Specifications

Density 1.0260g/mL
Boiling Point 195°C
Flash Point 83°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4130 to 1.415
Linear Formula (CH3O)2CHCH2CN
Quantity 100 mL
Specific Gravity 1.026
Synonym 3,3-dimethoxypropionitrile, propanenitrile, 3,3-dimethoxy, 3,3-dimethoxypropiononitrile, 3,3-dimethoxy propionitrile, 3,3-dimethoxy-propionitrile, 3,3-dimethoxy-propanenitrile, 3,3-dimethoxypropanitrile, acmc-209lz7, #, propanenitrile,3,3-dimethoxy
Solubility Information Solubility in water: soluble
Formula Weight 115.13
Percent Purity 90%
Chemical Name or Material 3, 3-Dimethoxypropanenitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doc
EINECSNumber 260-842-1

RUO – Research Use Only