missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethoxybenzil, 99+%

Catalog No. AAL1156406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1156406 5 g
AAL1156403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1156406 Supplier Thermo Scientific Chemicals Supplier No. L1156406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40101-17-5 | C16H14O4 | 270.284 g/mol

3,3'-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 40101-17-5
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00038221
InChI Key PJGXOGKIVAJFTE-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzil, 1,2-bis 3-methoxyphenyl ethane-1,2-dione, ethanedione, bis 3-methoxyphenyl, 1,2-ethanedione, 1,2-bis 3-methoxyphenyl, bis 3-methoxyphenyl ethane-1,2-dione, bis 3-methoxyphenyl ethanedione, acmc-20a2cz, 3,3'-dimethoxy benzil, d01vau, benzil-based compound, 22
PubChem CID 123493
IUPAC Name 1,2-bis(3-methoxyphenyl)ethane-1,2-dione
SMILES COC1=CC=CC(=C1)C(=O)C(=O)C2=CC(=CC=C2)OC

Specifications

Melting Point 81°C to 83°C
Quantity 5 g
Beilstein 2382124
Solubility Information Insoluble in water.
Formula Weight 270.29
Percent Purity ≥99%
Chemical Name or Material 3,3'-Dimethoxybenzil
EINECSNumber 254-793-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only