missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dihexyl-2,2'-bithiophene 98.0+%, TCI America™

Catalog No. D41801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41801G 1 g
D41805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41801G Supplier TCI America Supplier No. D41801G

Chemical Identifiers

CAS 125607-30-9
Molecular Formula C20H30S2
Molecular Weight (g/mol) 334.58
InChI Key RQHMOXZEVWCNMG-UHFFFAOYSA-N
PubChem CID 11484438
IUPAC Name 3-hexyl-2-(3-hexylthiophen-2-yl)thiophene
SMILES CCCCCCC1=C(SC=C1)C2=C(C=CS2)CCCCCC

Specifications

Color Yellow
Boiling Point 82°C
Quantity 1 g
Formula Weight 334.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3′-Dihexyl-2,2′-bithiophene
TSCA No
Recommended Storage Refrigerator