missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluoropyrrolidine Hydrochloride 98.0+%, TCI America™

Catalog No. D4315200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4315200MG 200 mg
D43151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4315200MG Supplier TCI America Supplier No. D4315200MG

Chemical Identifiers

CAS 163457-23-6
Molecular Formula C4H8ClF2N
Molecular Weight (g/mol) 143.562
MDL Number MFCD03788948
InChI Key YYVPZQADFREIFR-UHFFFAOYSA-N
Synonym 3,3-difluoropyrrolidine hydrochloride, 3,3-difluoropyrrolidine hcl, 3,3-difluoropyrrolidinehydrochloride, pyrrolidine, 3,3-difluoro-, hydrochloride, pyrrolidine, 3,3-difluoro-, hydrochloride 1:1, 3,3-difluoro-pyrrolidine hcl, 3,3-difluoropyrrolidine hcl salt, 3,3-difluoropyrrolidine, chloride, 3,3-difluoro-pyrrolidinehydrochloride, pubchem9358
PubChem CID 24903482
IUPAC Name 3,3-difluoropyrrolidine;hydrochloride
SMILES C1CNCC1(F)F.Cl

Specifications

Melting Point 134°C
Color Gray-White
Quantity 200 mg
Formula Weight 143.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Difluoropyrrolidine Hydrochloride