missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluoropyrrolidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. p-7047807
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19780ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL19780ME Supplier Thermo Scientific Chemicals Supplier No. L19780ME

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163457-23-6
Molecular Formula C4H8ClF2N
Molecular Weight (g/mol) 143.562
MDL Number MFCD03788948
InChI Key YYVPZQADFREIFR-UHFFFAOYSA-N
Synonym 3,3-difluoropyrrolidine hydrochloride, 3,3-difluoropyrrolidine hcl, 3,3-difluoropyrrolidinehydrochloride, pyrrolidine, 3,3-difluoro-, hydrochloride, pyrrolidine, 3,3-difluoro-, hydrochloride 1:1, 3,3-difluoro-pyrrolidine hcl, 3,3-difluoropyrrolidine hcl salt, 3,3-difluoropyrrolidine, chloride, 3,3-difluoro-pyrrolidinehydrochloride, pubchem9358
PubChem CID 24903482
IUPAC Name 3,3-difluoropyrrolidine;hydrochloride
SMILES C1CNCC1(F)F.Cl

Specifications

Melting Point 130°C to 134°C
Quantity 500 mg
Sensitivity Hygroscopic
Formula Weight 143.56
Percent Purity 98%
Chemical Name or Material 3,3-Difluoropyrrolidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only