missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluorocyclobutanecarboxylic Acid 98.0+%, TCI America™

Catalog No. D5225200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5225200MG 200 mg
D52251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5225200MG Supplier TCI America Supplier No. D5225200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107496-54-8
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.098
MDL Number MFCD08705857
InChI Key PLRCVBKYFLWAAT-UHFFFAOYSA-N
Synonym 3,3-difluorocyclobutanecarboxylic acid, 3,3-difluoro-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3,3-difluoro, cyclobutanecarboxylicacid, 3,3-difluoro, 3,3-difluorocyclobutanecarboxylicacid, acmc-209yhn, 3,3-difluorocyclopropyl methanol, 3,3-difluorocyclobutane carboxylic acid, 3,3-difluorocyclobutane-carboxylic acid, 3,3-difluoro-cyclobutane-1-carboxylic acid
PubChem CID 13777789
IUPAC Name 3,3-difluorocyclobutane-1-carboxylic acid
SMILES C1C(CC1(F)F)C(=O)O

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 205°C
Quantity 200 mg
UN Number 3261
Formula Weight 136.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Difluorocyclobutanecarboxylic Acid