missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Difluorobiphenyl, 97%

Catalog No. AAL1674003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1674003 1 g
AAL1674006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1674003 Supplier Thermo Scientific Chemicals Supplier No. L1674003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 396-64-5 | C12H8F2 | 190.19 g/mol

3,3'-Difluorobiphenyl is used materials in ferroelectric display devices

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Difluorobiphenyl is used materials in ferroelectric display devices

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 396-64-5
Molecular Formula C12H8F2
Molecular Weight (g/mol) 190.19
MDL Number MFCD00039216
InChI Key GAYJHUJLHJWCTH-UHFFFAOYSA-N
Synonym 3,3'-difluorobiphenyl, 3,3'-difluoro-1,1'-biphenyl, 1-fluoro-3-3-fluorophenyl benzene, 1,1'-biphenyl,3,3'-difluoro, 1,1'-biphenyl, 3,3'-difluoro, 3-fluoro-1-3-fluorophenyl benzene, acmc-1agdg, 3,3'-difluoro biphenyl, gayjhujlhjwcth-uhfffaoysa, 3,3'-difluoro-1,1'-biphenyl #
PubChem CID 123058
IUPAC Name 1-fluoro-3-(3-fluorophenyl)benzene
SMILES FC1=CC(=CC=C1)C1=CC(F)=CC=C1

Specifications

Melting Point 7°C to 8°C
Density 1.21
Boiling Point 130°C (14 mmHg)
Refractive Index 1.568
Quantity 1 g
UN Number UN3151
Beilstein 2046445
Solubility Information Difficult to mix.
Formula Weight 190.19
Percent Purity 97%
Chemical Name or Material 3,3'-Difluorobiphenyl
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, LIQUID
EINECSNumber 206-905-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only