missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluoroazetidine hydrochloride, 95%

Catalog No. p-7028431
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25751MD 250 mg
AAH2575103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25751MD Supplier Thermo Scientific Chemicals Supplier No. H25751MD
Only null left

CAS: 288315-03-7 | C3H6ClF2N | 129.535 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 288315-03-7
Molecular Formula C3H6ClF2N
MDL Number MFCD05663714
InChI Key CDBAEFXTCRKJPZ-UHFFFAOYSA-N
Synonym 3,3-difluoroazetidine hydrochloride, 3,3-difluoroazetidine hcl, azetidine, 3,3-difluoro-, hydrochloride, azetidine, 3,3-difluoro-, hydrochloride 1:1, 3,3-difluoroazetidine hydrochloride 1:1, 3,3-difluoroazetidinehydrochloride, acmc-1clfn, 3,3-difluoroazetidine-hcl, difluoroazetidine hydrochloride, 3,3-difluoroazetidin hydrochloride
PubChem CID 2758247
IUPAC Name 3,3-difluoroazetidine;hydrochloride

Specifications

Melting Point 138°C to 140°C
Quantity 250 mg
Sensitivity Hygroscopic
SMILES C1C(CN1)(F)F.Cl
Molecular Weight (g/mol) 129.535
Formula Weight 129.54
Percent Purity 95%
Chemical Name or Material 3,3-Difluoroazetidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only