missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethyloxadicarbocyanine iodide, 96%

Catalog No. p-7031858
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3309403 1 g
AAH3309406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3309403 Supplier Thermo Scientific Chemicals Supplier No. H3309403
Only null left

CAS: 14806-50-9 | C23H23IN2O2 | 486.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14806-50-9
Molecular Formula C23H23IN2O2
Molecular Weight (g/mol) 486.35
MDL Number MFCD00011953,MFCD00011953
InChI Key CLDZYSUDOQXJOU-UHFFFAOYSA-M
Synonym dodc iodide, 3-ethyl-2-5-3-ethyl-2-benzoxazolinylidene-1,3-pentadienyl benzoxazolium iodide, 5-3-ethylbenzoxazol-2-ylidene-1-3-ethylbenzoxazolium-2-yl-1,3-pentadiene iodide
PubChem CID 91972131
ChEBI CHEBI:51899
IUPAC Name (2Z)-3-ethyl-2-[5-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzoxazole;iodide
SMILES [I-].CCN1\C(OC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CC)C2=CC=CC=C2O1

Specifications

Melting Point ∽215°C (decomposition)
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 486.36
Percent Purity 96%
Chemical Name or Material 3,3'-Diethyloxadicarbocyanine iodide
Hazard Category H300
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 238-873-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only