missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diethoxypropionitrile 95.0+%, TCI America™

Catalog No. C117810ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C117810ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C117810ML Supplier TCI America Supplier No. C117810ML
Only null left

Chemical Identifiers

CAS 2032-34-0
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00040558
InChI Key WBOXEOCWOCJQNK-UHFFFAOYSA-N
Synonym 3,3-diethoxypropionitrile, 3,3-diethoxypropiononitrile, propanenitrile, 3,3-diethoxy, cyanoacetaldehyde diethyl acetal, 1,1-diethoxy-2-cyanoethane, 3,3-diethoxy propionitrile, cyadea, acmc-1cmza, 3,3-diethoxypropanitrile, 3,3-diethoxypropiononitril
PubChem CID 74851
IUPAC Name 3,3-diethoxypropanenitrile
SMILES CCOC(CC#N)OCC

Specifications

Color Yellow
Boiling Point 99°C
Quantity 10 mL
Formula Weight 143.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Diethoxypropionitrile