missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

$414.85 - $414.85

Chemical Identifiers

CAS 165904-27-8
Molecular Formula C13H27BO4
Molecular Weight (g/mol) 258.165
MDL Number MFCD03788723
InChI Key VEDSPUPKZZMMLL-UHFFFAOYSA-N
Synonym 3,3-diethoxy-1-propylboronic acid pinacol ester, 2-3,3-diethoxypropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-propanal diethyl acetal
PubChem CID 12115795
IUPAC Name 2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC(OCC)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1953203
View Documents
Thermo Scientific Chemicals
L1953203
1 g
Each for $414.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

165904-27-8
258.165
VEDSPUPKZZMMLL-UHFFFAOYSA-N
12115795
B1(OC(C(O1)(C)C)(C)C)CCC(OCC)OCC
C13H27BO4
MFCD03788723
3,3-diethoxy-1-propylboronic acid pinacol ester, 2-3,3-diethoxypropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-propanal diethyl acetal
2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

165904-27-8
1.428
1 g
VEDSPUPKZZMMLL-UHFFFAOYSA-N
2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
12115795
97%
C13H27BO4
MFCD03788723
3,3-diethoxy-1-propylboronic acid pinacol ester, 2-3,3-diethoxypropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-propanal diethyl acetal
B1(OC(C(O1)(C)C)(C)C)CCC(OCC)OCC
258.165
258.17
3,3-Diethoxy-1-propylboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only