missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. AAL1953203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1953203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1953203 Supplier Thermo Scientific Chemicals Supplier No. L1953203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 165904-27-8 | Molecular Formula: C13H27BO4 | Molecular Weight: 258.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 165904-27-8
Molecular Formula C13H27BO4
Molecular Weight (g/mol) 258.165
MDL Number MFCD03788723
InChI Key VEDSPUPKZZMMLL-UHFFFAOYSA-N
Synonym 3,3-diethoxy-1-propylboronic acid pinacol ester, 2-3,3-diethoxypropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-propanal diethyl acetal
PubChem CID 12115795
IUPAC Name 2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC(OCC)OCC

Specifications

Refractive Index 1.428
Quantity 1 g
Formula Weight 258.17
Percent Purity 97%
Chemical Name or Material 3,3-Diethoxy-1-propylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only