missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dibromo-4,4'-biphenol 98.0+%, TCI America™

$428.73 - $1403.63

Chemical Identifiers

CAS 189039-64-3
Molecular Formula C12H8Br2O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D36021G
View Documents
TCI America
D36021G
1 g
Each for $428.73
Only null left
 
D36025G
View Documents
TCI America
D36025G
5 g
Each for $1,403.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

189039-64-3
C12H8Br2O2

Specifications

189039-64-3
White-Yellow
1 g
344.00
Crystalline Powder
122°C
C12H8Br2O2
3,3′C-Dibromobiphenyl-4,4′C-diol, 3,3′C-Dibromo-4,4′C-dihydroxybiphenyl
≥98.0% (T)
3,3′-Dibromo-4,4′-biphenol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'