missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dibromo-4,4'-biphenol 98.0+%, TCI America™

Catalog No. D36025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36025G 5 g
D36021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36025G Supplier TCI America Supplier No. D36025G
Only null left

Chemical Identifiers

CAS 189039-64-3
Molecular Formula C12H8Br2O2

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 5 g
Synonym 3,3′-Dibromobiphenyl-4,4′-diol, 3,3′-Dibromo-4,4′-dihydroxybiphenyl
Formula Weight 344.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dibromo-4,4′-biphenol