missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminodipropylamine 98.0+%, TCI America™

Catalog No. p-7126022
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D009025ML 25 mL
D0090500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D009025ML Supplier TCI America Supplier No. D009025ML
Only null left

Chemical Identifiers

CAS 56-18-8
Molecular Formula C6H17N3
Molecular Weight (g/mol) 131.223
MDL Number MFCD00008214
InChI Key OTBHHUPVCYLGQO-UHFFFAOYSA-N
Synonym norspermidine, bis 3-aminopropyl amine, dipropylenetriamine, 3,3'-diaminodipropylamine, 3,3'-iminobispropylamine, caldine, 3,3'-iminobis propylamine, 1,7-diamino-4-azaheptane, 4-azaheptane-1,7-diamine, 3,3-diaminodipropylamine
PubChem CID 5942
ChEBI CHEBI:16841
IUPAC Name N'-(3-aminopropyl)propane-1,3-diamine
SMILES C(CN)CNCCCN

Specifications

Melting Point -14°C
Color Yellow
pH 12 (100g/L H2O soln. 20°C)
Boiling Point 115°C
Quantity 25 mL
UN Number 2269
Formula Weight 131.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,3′-Diaminodipropylamine
EINECSNumber (2)-0148
RTECSNumber JL9450000
TSCA Yes