missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Diaminodipropylamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -14°C |
| Color | Yellow |
| pH | 12 (100g/L H2O soln. 20°C) |
| Boiling Point | 115°C |
| Quantity | 500 mL |
| UN Number | 2269 |
| Formula Weight | 131.22 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3,3′-Diaminodipropylamine |
Safety and Handling
| EINECSNumber | (2)-0148 |
| RTECSNumber | JL9450000 |
| TSCA | Yes |