missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Diaminobenzidine 98.0+%, TCI America™
Supplier: TCI America D007725G
Specifications
| 3,3′-Diaminobenzidine | |
| 177°C | |
| MFCD00007725 | |
| 3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine | |
| C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N | |
| 214.272 | |
| 214.27 | |
| Crystalline Powder |
| 91-95-2 | |
| C12H14N4 | |
| 25 g | |
| HSTOKWSFWGCZMH-UHFFFAOYSA-N | |
| 4-(3,4-diaminophenyl)benzene-1,2-diamine | |
| 7071 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 91-95-2 | |
| 214.272 | |
| HSTOKWSFWGCZMH-UHFFFAOYSA-N | |
| 7071 | |
| C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N |
| C12H14N4 | |
| MFCD00007725 | |
| 3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine | |
| 4-(3,4-diaminophenyl)benzene-1,2-diamine |
Safety and Handling
EINECSNumber : (4)-1475
RTECSNumber : DV8750000
TSCA : Yes
Recommended Storage : Refrigerator