missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Diaminobenzidine 97.0+%, TCI America™
$44.28 - $132.84
Chemical Identifiers
| CAS | 91-95-2 |
|---|---|
| Molecular Formula | C12H14N4 |
| Molecular Weight (g/mol) | 214.272 |
| MDL Number | MFCD00007725 |
| InChI Key | HSTOKWSFWGCZMH-UHFFFAOYSA-N |
| Synonym | 3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine |
| PubChem CID | 7071 |
| IUPAC Name | 4-(3,4-diaminophenyl)benzene-1,2-diamine |
| SMILES | C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N |
Chemical Identifiers
| 91-95-2 | |
| 214.272 | |
| HSTOKWSFWGCZMH-UHFFFAOYSA-N | |
| 7071 | |
| C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N |
| C12H14N4 | |
| MFCD00007725 | |
| 3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine | |
| 4-(3,4-diaminophenyl)benzene-1,2-diamine |
Specifications
| 91-95-2 | |
| C12H14N4 | |
| 1 g | |
| HSTOKWSFWGCZMH-UHFFFAOYSA-N | |
| 4-(3,4-diaminophenyl)benzene-1,2-diamine | |
| 7071 | |
| ≥97.0% (T) | |
| 3,3′-Diaminobenzidine [for Biochemical Research] |
| 177°C | |
| MFCD00007725 | |
| 3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine | |
| C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N | |
| 214.272 | |
| 214.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-1475
RTECSNumber : DV8750000
TSCA : Yes
Recommended Storage : Refrigerator