missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine 97.0+%, TCI America™

Catalog No. D37561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37561G 1 g
D37565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37561G Supplier TCI America Supplier No. D37561G

Chemical Identifiers

CAS 91-95-2
Molecular Formula C12H14N4
Molecular Weight (g/mol) 214.272
MDL Number MFCD00007725
InChI Key HSTOKWSFWGCZMH-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine
PubChem CID 7071
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine
SMILES C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N

Specifications

Melting Point 177°C
Quantity 1 g
Formula Weight 214.27
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diaminobenzidine [for Biochemical Research]
EINECSNumber (4)-1475
RTECSNumber DV8750000
TSCA Yes
Recommended Storage Refrigerator