missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Diamino-N-methyldipropylamine 98.0+%, TCI America™
$51.98 - $225.25
Chemical Identifiers
| CAS | 105-83-9 |
|---|---|
| Molecular Formula | C7H19N3 |
| Molecular Weight (g/mol) | 145.25 |
| MDL Number | MFCD00008217 |
| InChI Key | KMBPCQSCMCEPMU-UHFFFAOYSA-N |
| Synonym | n,n-bis 3-aminopropyl methylamine, 3,3'-diamino-n-methyldipropylamine, bis 3-aminopropyl methylamine, n-methyliminobis propylamine, methyliminobispropylamine, methylbis 3-aminopropyl amine, 5-methyldipropylenetriamine, 1,3-propanediamine, n-3-aminopropyl-n-methyl, 3,3'-methyliminobispropylamine, n1-3-aminopropyl-n1-methylpropane-1,3-diamine |
| PubChem CID | 7777 |
| IUPAC Name | bis(3-aminopropyl)(methyl)amine |
| SMILES | CN(CCCN)CCCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B082125ML
|
TCI America
B082125ML |
25 mL |
Each for $51.98
|
|
|||||
|
B0821500ML
|
TCI America
B0821500ML |
500 mL |
Each for $225.25
|
|
|||||
Chemical Identifiers
| 105-83-9 | |
| 145.25 | |
| KMBPCQSCMCEPMU-UHFFFAOYSA-N | |
| 7777 | |
| CN(CCCN)CCCN |
| C7H19N3 | |
| MFCD00008217 | |
| n,n-bis 3-aminopropyl methylamine, 3,3'-diamino-n-methyldipropylamine, bis 3-aminopropyl methylamine, n-methyliminobis propylamine, methyliminobispropylamine, methylbis 3-aminopropyl amine, 5-methyldipropylenetriamine, 1,3-propanediamine, n-3-aminopropyl-n-methyl, 3,3'-methyliminobispropylamine, n1-3-aminopropyl-n1-methylpropane-1,3-diamine | |
| bis(3-aminopropyl)(methyl)amine |
Specifications
| 105-83-9 | |
| 241°C | |
| MFCD00008217 | |
| 2927 | |
| KMBPCQSCMCEPMU-UHFFFAOYSA-N | |
| bis(3-aminopropyl)(methyl)amine | |
| 7777 | |
| ≥98.0% (GC,T) | |
| 3,3′-Diamino-N-methyldipropylamine |
| Colorless | |
| C7H19N3 | |
| 25 mL | |
| n,n-bis 3-aminopropyl methylamine, 3,3'-diamino-n-methyldipropylamine, bis 3-aminopropyl methylamine, n-methyliminobis propylamine, methyliminobispropylamine, methylbis 3-aminopropyl amine, 5-methyldipropylenetriamine, 1,3-propanediamine, n-3-aminopropyl-n-methyl, 3,3'-methyliminobispropylamine, n1-3-aminopropyl-n1-methylpropane-1,3-diamine | |
| CN(CCCN)CCCN | |
| 145.25 | |
| 145.25 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0165
RTECSNumber : JL9625000
TSCA : Yes