missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392

Catalog No. p-7080097
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4640314 25 g
AA4640322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4640314 Supplier Thermo Scientific Chemicals Supplier No. 04640314

CAS: 1071466-61-9 | C25H28O4 | 392.495 g/mol

It is used in epoxy systems or in combination with acrylates, methacrylates, and maleimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Recommended for use in epoxy systems or in combination with acrylates, methacrylates, and maleimides

Chemical Identifiers

CAS 1071466-61-9
Molecular Formula C25H28O4
Molecular Weight (g/mol) 392.495
MDL Number MFCD22200539
InChI Key GJTZQXNRNNDGRG-UHFFFAOYSA-N
Synonym 2,2'-diallyl bisphenol a diacetate ether, 4,4'-isopropylidene bis 2-allylphenol acetate, phenyl ester epoxy curative hybrid of bisphenol a, 3,3'-diallylbisphenol a diacetate, epoxy curative, ec-392, 4-2-4-acetyloxy-3-prop-2-en-1-yl phenyl propan-2-yl-2-prop-2-en-1-yl phenyl acetate
PubChem CID 59215472
IUPAC Name [4-[2-(4-acetyloxy-3-prop-2-enylphenyl)propan-2-yl]-2-prop-2-enylphenyl] acetate
SMILES CC(=O)OC1=C(C=C(C=C1)C(C)(C)C2=CC(=C(C=C2)OC(=O)C)CC=C)CC=C

Specifications

Color Amber
Physical Form Amber liquid
Quantity 25 g
Formula Weight 392.5
Chemical Name or Material 3,3'-Diallylbisphenol A diacetate, Epoxy curative, EC-392
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only