missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Di-n-propylthiadicarbocyanine iodide, 96%

Catalog No. p-7032658
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3420903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3420903 Supplier Thermo Scientific Chemicals Supplier No. H3420903
Only null left

CAS: 53213-94-8 | C25H27IN2S2 | 546.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53213-94-8
Molecular Formula C25H27IN2S2
Molecular Weight (g/mol) 546.53
MDL Number MFCD00075033,MFCD00075033
InChI Key GDEURKKLNUGTDA-UHFFFAOYSA-M
Synonym disc3 5, 3,3'-dipropylthiadicarbocyanine iodide, 3-propyl-2-5-3-propyl-2 3h-benzothiazolylidene-1,3-pentadienyl benzothiazolium iodide, 5-3-propylbenzothiazol-2-ylidene-1-3-propylbenzothiazolium-2-yl-1,3-pentadiene iodide
PubChem CID 91972132
SMILES [I-].CCCN1\C(SC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CCC)C2=CC=CC=C2S1

Specifications

Melting Point >250°C
Quantity 1 g
Sensitivity Light sensitive
IUPAC Name 3-propyl-2-[(3E)-5-[(2Z)-3-propyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
Formula Weight 546.54
Percent Purity 96%
Chemical Name or Material 3,3'-Di-n-propylthiadicarbocyanine iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only