missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)pyridine 98.0+%, TCI America™

Catalog No. B48651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48651G 1 g
B48655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48651G Supplier TCI America Supplier No. B48651G
Only null left

Chemical Identifiers

CAS 4422-32-6
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD16658917
InChI Key JHUIVUBQMBPYJE-UHFFFAOYSA-N
PubChem CID 13150401
IUPAC Name 3-(3-bromophenyl)pyridine
SMILES C1=CC(=CC(=C1)Br)C2=CN=CC=C2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(3-Bromophenyl)pyridine