missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[3-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 250mg |
| Formula Weight | 253.1 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-[3-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole |