missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Bis[di(p-tolyl)amino]biphenyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 193°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 544.74 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,3′-Bis[di(p-tolyl)amino]biphenyl |
Safety and Handling
| TSCA | No |