missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Benzyloxy)phenyl]propionic acid, 96%

Catalog No. AAH3389906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3389906 5 g
AAH3389903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3389906 Supplier Thermo Scientific Chemicals Supplier No. H3389906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57668-34-5 | C16H16O3 | 256.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57668-34-5
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD09258905
InChI Key HVXOYUOKPLHWNU-UHFFFAOYSA-N
Synonym 3-3-benzyloxy phenyl propanoic acid, benzenepropanoic acid, 3-phenylmethoxy, 3-3-benzyloxy-phenyl-propanoic acid, 3-3-benzyloxy phenyl propionic acid, 3-3-phenylmethyl oxy phenyl propanoic acid
PubChem CID 21938504
IUPAC Name 3-(3-phenylmethoxyphenyl)propanoic acid
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)CCC(=O)O

Specifications

Melting Point 83°C to 85°C
Quantity 5 g
Formula Weight 256.3
Percent Purity 96%
Chemical Name or Material 3-[3-(Benzyloxy)phenyl]propionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only