missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',5-Triiodo-L-thyronine 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 100 mg |
| Formula Weight | 650.98 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,3′,5-Triiodo-L-thyronine |
Safety and Handling
| RTECSNumber | AY6750000 |
| TSCA | No |