missing translation for 'onlineSavingsMsg'
Learn More

3-(3,5-Dibromophenyl)propionic acid, 97%

Catalog No. p-7035804
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5401203 1 g
AAH5401206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5401203 Supplier Thermo Scientific Chemicals Supplier No. H5401203
Only null left

CAS: 923977-15-5 | C9H8Br2O2 | 307.969 g/mol

3-(3,5-Dibromophenyl)propionic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3,5-Dibromophenyl)propionic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 923977-15-5
Molecular Formula C9H8Br2O2
Molecular Weight (g/mol) 307.969
MDL Number MFCD11973898
InChI Key AMKWPMUHANVQSZ-UHFFFAOYSA-N
Synonym 3-3,5-dibromophenyl propanoic acid, 3-3,5-dibromophenyl propionic acid, 3,5-dibromobenzenepropanoic acid, 3-3,5-dibromo-phenyl-propionic acid
PubChem CID 16038256
IUPAC Name 3-(3,5-dibromophenyl)propanoic acid
SMILES C1=C(C=C(C=C1Br)Br)CCC(=O)O

Specifications

Melting Point 62°C
Quantity 1 g
Beilstein 10537616
Formula Weight 307.97
Percent Purity 97%
Chemical Name or Material 3-(3,5-Dibromophenyl)propionic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only