missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich 3,3',5,5'-Tetramethylbenzidine
Description
3,3',5,5'-Tetramethylbenzidine
Spécifications
Spécifications
| CAS | 54827-17-7 |
| Melting Point | 168°C to 171°C (lit.) |
| Molecular Formula | C16H20N2 |
| Linear Formula | [-C6H2(CH3)2-4-NH2]2 |
| Quantity | 25 g |
| MDL Number | MFCD00007748 |
| Synonym | BM blue; Sure Blue TMB; TMB; TMB Blotting Plus; TMB substrate |
| Molecular Weight (g/mol) | 240.34 |
| Percent Purity | ≥99% |