missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dimethoxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. D195310G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D195310G 10 g
D195325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D195310G Supplier TCI America Supplier No. D195310G

Chemical Identifiers

CAS 2107-70-2
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00002774
InChI Key LHHKQWQTBCTDQM-UHFFFAOYSA-N
Synonym 3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl propanoic acid, 3,4-dimethoxyhydrocinnamic acid, benzenepropanoic acid, 3,4-dimethoxy, 3-3,4-dimethoxyphenyl-propionic acid, 3-3,4-dimethoxy-phenyl-propionic acid, unii-6xo32zsp1d, 6xo32zsp1d, 3-3',4'-dimethoxyphenyl propanoic acid, 3-3,4-dimethoxyphenyl propionicacid
PubChem CID 75019
ChEBI CHEBI:44235
IUPAC Name 3-(3,4-dimethoxyphenyl)propanoic acid
SMILES COC1=C(C=C(C=C1)CCC(=O)O)OC

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 10 g
Formula Weight 210.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3,4-Dimethoxyphenyl)propionic Acid