missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dimethoxyphenyl)propionic acid, 98%

Catalog No. AAA1432506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1432506 5 g
AAA1432514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1432506 Supplier Thermo Scientific Chemicals Supplier No. A1432506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2107-70-2 | C11H14O4 | 210.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2107-70-2
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00002774
InChI Key LHHKQWQTBCTDQM-UHFFFAOYSA-N
Synonym 3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl propanoic acid, 3,4-dimethoxyhydrocinnamic acid, benzenepropanoic acid, 3,4-dimethoxy, 3-3,4-dimethoxyphenyl-propionic acid, 3-3,4-dimethoxy-phenyl-propionic acid, unii-6xo32zsp1d, 6xo32zsp1d, 3-3',4'-dimethoxyphenyl propanoic acid, 3-3,4-dimethoxyphenyl propionicacid
PubChem CID 75019
ChEBI CHEBI:44235
IUPAC Name 3-(3,4-dimethoxyphenyl)propanoic acid
SMILES COC1=C(C=C(C=C1)CCC(=O)O)OC

Specifications

Melting Point 96°C to 99°C
Quantity 5 g
Beilstein 2696272
Formula Weight 210.23
Percent Purity 98%
Chemical Name or Material 3-(3,4-Dimethoxyphenyl)propionic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 218-288-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only