missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dihydroxyphenyl)-L-alanine 98.0+%, TCI America™

Catalog No. p-7126313
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06005G 5 g
D060025G 25 g
D0600100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D06005G Supplier TCI America Supplier No. D06005G
Only null left

Chemical Identifiers

CAS 59-92-7
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00002598
InChI Key WTDRDQBEARUVNC-LURJTMIESA-N
Synonym levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
PubChem CID 6047
ChEBI CHEBI:15765
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

Melting Point 278°C
Color White
Quantity 5 g
Formula Weight 197.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3,4-Dihydroxyphenyl)-L-alanine
EINECSNumber (9)-1555
RTECSNumber AY5600000
TSCA Yes