missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dihydroxyphenyl)-L-alanine, 99%

3-(3,4-Dihydroxyphenyl)-L-alanine (levodopa orL-dopa), CAS # 59-92-7, is a dopamine precursor molecule. | CAS: 59-92-7 | C9H11NO4 | 197.19 g/mol

Supplier:  Thermo Scientific Chemicals 167530250

 View more versions of this product

Catalog No. AC167530250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 3-(3,4-Dihydroxyphenyl)-L-alanine, or L-dopa, is converted to dopamine by DOPA decarboxylase in the body

• Upon conversion to dopamine, it acts as an excitatory neurotransmitter

Applications

• L-dopa is useful in the laboratory study of Parkinson’s to replenish dopamine in depleted circuits

• L-dopa has also been used for laboratory research on hypertension and melanocyte activity

Specifications

Specifications

3-(3, 4-Dihydroxyphenyl)-L-alanine
Cream to White
1% max.
C9H11NO4
(HO)2C6H3CH2CH(NH2)CO2H
14, IV, 2492
0.1% max.
WTDRDQBEARUVNC-LURJTMIESA-N
(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
25 g
CHEBI:15765
≥98.5%
59-92-7
Authentic
99%
Glass Bottle
MFCD00002598
15, 5516
levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
197.19
6047
197.19
Crystalline Powder

Chemical Identifiers

59-92-7
197.19
WTDRDQBEARUVNC-LURJTMIESA-N
6047
(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
C9H11NO4
MFCD00002598
levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
CHEBI:15765
N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only