missing translation for 'onlineSavingsMsg'
Learn More

3,3,4,4-Tetrafluoropyrrolidine Hydrochloride 98.0+%, TCI America™

Catalog No. T2342100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2342100MG 100 mg
T23421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2342100MG Supplier TCI America Supplier No. T2342100MG

Chemical Identifiers

CAS 1810-13-5
Molecular Formula C4H6ClF4N
Molecular Weight (g/mol) 179.54
MDL Number MFCD06200835
InChI Key KPTMFLKIYHWQPB-UHFFFAOYSA-N
PubChem CID 22645920
IUPAC Name 3,3,4,4-tetrafluoropyrrolidine hydrochloride
SMILES Cl.FC1(F)CNCC1(F)F

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 179.54
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,4,4-Tetrafluoropyrrolidine Hydrochloride