missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3,3-Trifluoropropionic Acid 98.0+%, TCI America™
$50.33 - $137.94
Chemical Identifiers
| CAS | 2516-99-6 |
|---|---|
| Molecular Formula | C3H3F3O2 |
| Molecular Weight (g/mol) | 128.05 |
| MDL Number | MFCD00153292 |
| InChI Key | KSNKQSPJFRQSEI-UHFFFAOYSA-N |
| Synonym | 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid |
| PubChem CID | 2777972 |
| IUPAC Name | 3,3,3-trifluoropropanoic acid |
| SMILES | C(C(=O)O)C(F)(F)F |
Chemical Identifiers
| 2516-99-6 | |
| 128.05 | |
| KSNKQSPJFRQSEI-UHFFFAOYSA-N | |
| 2777972 | |
| C(C(=O)O)C(F)(F)F |
| C3H3F3O2 | |
| MFCD00153292 | |
| 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid | |
| 3,3,3-trifluoropropanoic acid |
Specifications
| 2516-99-6 | |
| 153°C | |
| MFCD00153292 | |
| 2924 | |
| KSNKQSPJFRQSEI-UHFFFAOYSA-N | |
| 3,3,3-trifluoropropanoic acid | |
| 2777972 | |
| ≥98.0% (GC,T) | |
| 3,3,3-Trifluoropropionic Acid |
| Yellow | |
| C3H3F3O2 | |
| 1 g | |
| 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid | |
| C(C(=O)O)C(F)(F)F | |
| 128.05 | |
| 128.05 | |
| Liquid |
Safety and Handling
TSCA : No