Learn More
3,3,3-Trifluoropropionic acid, 98%
CAS: 2516-99-6 | C3H3F3O2 | 128.05 g/mol
$771.44 - $771.44
Chemical Identifiers
| CAS | 2516-99-6 |
|---|---|
| Molecular Formula | C3H3F3O2 |
| Molecular Weight (g/mol) | 128.05 |
| MDL Number | MFCD00153292 |
| InChI Key | KSNKQSPJFRQSEI-UHFFFAOYSA-N |
| Synonym | 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid |
| PubChem CID | 2777972 |
| IUPAC Name | 3,3,3-trifluoropropanoic acid |
| SMILES | C(C(=O)O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2042814
|
Thermo Scientific Chemicals
B2042814 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2516-99-6 | |
| 128.05 | |
| KSNKQSPJFRQSEI-UHFFFAOYSA-N | |
| 2777972 | |
| C(C(=O)O)C(F)(F)F |
| C3H3F3O2 | |
| MFCD00153292 | |
| 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid | |
| 3,3,3-trifluoropropanoic acid |
Specifications
| 2516-99-6 | |
| 1.441 | |
| >100°C (212°F) | |
| 1.334 | |
| 25 g | |
| 1751796 | |
| KSNKQSPJFRQSEI-UHFFFAOYSA-N | |
| 3,3,3-trifluoropropanoic acid | |
| 2777972 | |
| 98% |
| 9°C | |
| 146°C | |
| C3H3F3O2 | |
| MFCD00153292 | |
| UN3265 | |
| 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid | |
| C(C(=O)O)C(F)(F)F | |
| 128.05 | |
| 128.05 | |
| 3,3,3-Trifluoropropionic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P374-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only