missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic Acid 98.0+%, TCI America™

Catalog No. T25871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25871G 1 g
T25875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25871G Supplier TCI America Supplier No. T25871G

Chemical Identifiers

CAS 662-22-6
Molecular Formula C4H2F6O3
Molecular Weight (g/mol) 212.047
MDL Number MFCD00190631
InChI Key CMQUGOHGJUTDGZ-UHFFFAOYSA-N
Synonym 2,2-bis trifluoromethyl-2-hydroxyacetic acid, 3,3,3-trifluoro-2-hydroxy-2-trifluoromethyl propanoic acid, 2,2-bis trifluoromethyl glycolic acid, 3,3,3-trifluoro-2-hydroxy-2-trifluoromethyl propionic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy-2-trifluoromethyl, acmc-1b1xb, alpha-hydroxyperfluoroisobutyric acid, 2,2-bis-trifluoromethyl 2-hydroxyacetic acid, 2,2-bis-trifluoromethyl-2-hydroxyacetic acid, 2,2-bis-trifluoromethyl-2-hydroxy-acetic acid
PubChem CID 2725043
IUPAC Name 3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanoic acid
SMILES C(=O)(C(C(F)(F)F)(C(F)(F)F)O)O

Specifications

Melting Point 84°C
Color White-Yellow
Boiling Point 153°C
Quantity 1 g
UN Number 3261
Formula Weight 212.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic Acid