missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-2-hydroxy-2-methylpropionic Acid 97.0+%, TCI America™

Catalog No. p-7137013
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19021G 1 g
T19025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19021G Supplier TCI America Supplier No. T19021G
Only null left

Chemical Identifiers

CAS 374-35-6
Molecular Formula C4H5F3O3
Molecular Weight (g/mol) 158.08
MDL Number MFCD00190646,MFCD03095400,MFCD07375449
InChI Key CTGJACFEVDCYMC-UHFFFAOYNA-N
Synonym 2-Hydroxy-2-(trifluoromethyl)propionic Acid, 3,3,3-Trifluoro-2-hydroxyisobutyric Acid
PubChem CID 2775125
IUPAC Name 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
SMILES CC(O)(C(O)=O)C(F)(F)F

Specifications

Melting Point 88°C
Color White
Boiling Point 189°C
Quantity 1 g
UN Number 3261
Formula Weight 158.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,3-Trifluoro-2-hydroxy-2-methylpropionic Acid
TSCA No
Recommended Storage Refrigerator