missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-1-propanol 98.0+%, TCI America™

Catalog No. T29861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29861G 1 g
T29865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29861G Supplier TCI America Supplier No. T29861G

Chemical Identifiers

CAS 2240-88-2
Molecular Formula C3H5F3O
Molecular Weight (g/mol) 114.067
MDL Number MFCD00190649
InChI Key HDBGBTNNPRCVND-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-1-propanol, 3,3,3-trifluoropropanol, 1-propanol, 3,3,3-trifluoro, trifluoropropanol, acmc-1cqb1, 3,3,3-trifluoropropanol-1, 2-trifluoromethyl ethan-1-ol, 3,3,3-trifluoro-propan-1-ol, poly difluoromethylene , alpha-fluoro-omega-2-hydroxyethyl, 3-hydroxy-1,1,1-trifluoropropane
PubChem CID 137512
IUPAC Name 3,3,3-trifluoropropan-1-ol
SMILES C(CO)C(F)(F)F

Specifications

Color Colorless
Boiling Point 100°C
Quantity 1 g
UN Number 1987
Formula Weight 114.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3,3-Trifluoro-1-propanol