missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-1-propanol, 97%

Catalog No. p-7058462
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAL1687903 1 g
AAL1687906 5 g
2 options

Catalog No. AAL1687903

Supplier: Thermo Scientific Chemicals L1687903

Only null left

CAS: 2240-88-2 | C3H5F3O | 114.067 g/mol

3,3,3-Trifluoro-1-propanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3,3-Trifluoro-1-propanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2240-88-2
Molecular Formula C3H5F3O
Molecular Weight (g/mol) 114.067
MDL Number MFCD00190649
InChI Key HDBGBTNNPRCVND-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-1-propanol, 3,3,3-trifluoropropanol, 1-propanol, 3,3,3-trifluoro, trifluoropropanol, acmc-1cqb1, 3,3,3-trifluoropropanol-1, 2-trifluoromethyl ethan-1-ol, 3,3,3-trifluoro-propan-1-ol, poly difluoromethylene , alpha-fluoro-omega-2-hydroxyethyl, 3-hydroxy-1,1,1-trifluoropropane
PubChem CID 137512
IUPAC Name 3,3,3-trifluoropropan-1-ol
SMILES C(CO)C(F)(F)F

Specifications

Density 1.294
Boiling Point 100°C to 101°C
Flash Point 37°C (98°F)
Refractive Index 1.32
Quantity 1 g
UN Number UN1987
Beilstein 1734777
Solubility Information Fully miscible in water.
Formula Weight 114.07
Percent Purity 97%
Chemical Name or Material 3,3,3-Trifluoro-1-propanol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only