missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-1-propanesulfonyl chloride, 95%, Thermo Scientific Chemicals

$723.78 - $723.78

Chemical Identifiers

CAS 845866-80-0
Molecular Formula C3H4ClF3O2S
Molecular Weight (g/mol) 196.568
MDL Number MFCD06248898
InChI Key OXKHPRFGCFJUSK-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-1-propanesulfonyl chloride, 3,3,3-tris fluoranyl propane-1-sulfonyl chloride, chloro 3,3,3-trifluoropropyl sulfone, 3,3,3-trifluoropropylsulfonyl chloride, 3,3,3-trifluoropropanesulfonyl chloride, 3,3,3-trifluoropropylsulphonyl chloride, 3,3,3-trifluoropropane sulfonyl chloride, 3,3,3-trifluoropropane-1-sulphonyl chloride, 1-propanesulfonylchloride, 3,3,3-trifluoro, 3,3,3-trifluoro-1-propane sulfonyl chloride
PubChem CID 2761177
IUPAC Name 3,3,3-trifluoropropane-1-sulfonyl chloride
SMILES C(CS(=O)(=O)Cl)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH25856ME
View Documents
Thermo Scientific Chemicals
H25856ME
500 mg
Each for $723.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

845866-80-0
196.568
OXKHPRFGCFJUSK-UHFFFAOYSA-N
2761177
C(CS(=O)(=O)Cl)C(F)(F)F
C3H4ClF3O2S
MFCD06248898
3,3,3-trifluoro-1-propanesulfonyl chloride, 3,3,3-tris fluoranyl propane-1-sulfonyl chloride, chloro 3,3,3-trifluoropropyl sulfone, 3,3,3-trifluoropropylsulfonyl chloride, 3,3,3-trifluoropropanesulfonyl chloride, 3,3,3-trifluoropropylsulphonyl chloride, 3,3,3-trifluoropropane sulfonyl chloride, 3,3,3-trifluoropropane-1-sulphonyl chloride, 1-propanesulfonylchloride, 3,3,3-trifluoro, 3,3,3-trifluoro-1-propane sulfonyl chloride
3,3,3-trifluoropropane-1-sulfonyl chloride

Specifications

845866-80-0
C3H4ClF3O2S
1.3997
500 mg
Air and moisture sensitive
OXKHPRFGCFJUSK-UHFFFAOYSA-N
3,3,3-trifluoropropane-1-sulfonyl chloride
2761177
95%
77°C to 78°C (16 mmHg)
CF3(CH2)2SO2Cl
MFCD06248898
UN3265
3,3,3-trifluoro-1-propanesulfonyl chloride, 3,3,3-tris fluoranyl propane-1-sulfonyl chloride, chloro 3,3,3-trifluoropropyl sulfone, 3,3,3-trifluoropropylsulfonyl chloride, 3,3,3-trifluoropropanesulfonyl chloride, 3,3,3-trifluoropropylsulphonyl chloride, 3,3,3-trifluoropropane sulfonyl chloride, 3,3,3-trifluoropropane-1-sulphonyl chloride, 1-propanesulfonylchloride, 3,3,3-trifluoro, 3,3,3-trifluoro-1-propane sulfonyl chloride
C(CS(=O)(=O)Cl)C(F)(F)F
196.568
196.57
3,3,3-Trifluoro-1-propanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only