missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',3″-Nitrilotripropionic Acid 99.0+%, TCI America™
$304.18 - $583.33
Chemical Identifiers
| CAS | 817-11-8 |
|---|---|
| Molecular Formula | C9H15NO6 |
| Molecular Weight (g/mol) | 233.22 |
| MDL Number | MFCD00055827 |
| InChI Key | IWTIBPIVCKUAHK-UHFFFAOYSA-N |
| Synonym | 3,3',3-nitrilotripropionic acid, nitrilotripropionic acid, 3,3',3-nitrilotripropanoic acid, beta-alanine, n,n-bis 2-carboxyethyl, 3-bis 2-carboxyethyl amino propanoic acid, acmc-209pm3, 3,3',3-nitrilotripropanoic acid non-preferred name, tris 3-oxo-3-hydroxypropyl amine |
| PubChem CID | 98537 |
| IUPAC Name | 3-[bis(2-carboxyethyl)amino]propanoic acid |
| SMILES | C(CN(CCC(=O)O)CCC(=O)O)C(=O)O |
Chemical Identifiers
| 817-11-8 | |
| 233.22 | |
| IWTIBPIVCKUAHK-UHFFFAOYSA-N | |
| 98537 | |
| C(CN(CCC(=O)O)CCC(=O)O)C(=O)O |
| C9H15NO6 | |
| MFCD00055827 | |
| 3,3',3-nitrilotripropionic acid, nitrilotripropionic acid, 3,3',3-nitrilotripropanoic acid, beta-alanine, n,n-bis 2-carboxyethyl, 3-bis 2-carboxyethyl amino propanoic acid, acmc-209pm3, 3,3',3-nitrilotripropanoic acid non-preferred name, tris 3-oxo-3-hydroxypropyl amine | |
| 3-[bis(2-carboxyethyl)amino]propanoic acid |
Specifications
| 817-11-8 | |
| C9H15NO6 | |
| 10 g | |
| IWTIBPIVCKUAHK-UHFFFAOYSA-N | |
| 3-[bis(2-carboxyethyl)amino]propanoic acid | |
| 98537 | |
| ≥99.0% (T) | |
| 3,3′,3′′-Nitrilotripropionic Acid |
| White | |
| MFCD00055827 | |
| 3,3',3-nitrilotripropionic acid, nitrilotripropionic acid, 3,3',3-nitrilotripropanoic acid, beta-alanine, n,n-bis 2-carboxyethyl, 3-bis 2-carboxyethyl amino propanoic acid, acmc-209pm3, 3,3',3-nitrilotripropanoic acid non-preferred name, tris 3-oxo-3-hydroxypropyl amine | |
| C(CN(CCC(=O)O)CCC(=O)O)C(=O)O | |
| 233.22 | |
| 233.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No