missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester, 97%

CAS: 915402-03-8 | C17H25BO2Si | 300.28 g/mol

$130.63 - $437.79

Chemical Identifiers

CAS 915402-03-8
Molecular Formula C17H25BO2Si
Molecular Weight (g/mol) 300.28
MDL Number MFCD08706285
InChI Key LDKJXCOLGXLWKP-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-3-trimethylsilanylethynyl-phenyl-1,3,2 dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl-trimethylsilane, trimethyl 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, 3-trimethylsilyl ethynyl phenylboronic acid pinacol ester, trimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, amtb201, 3-2-trimethylsilyl ethynyl benzeneboronic acid pinacol ester
PubChem CID 17999151
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C#C[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5873103
View Documents
Thermo Scientific Chemicals
H5873103
1 g
Each for $130.63
Only null left
 
AAH5873106
View Documents
Thermo Scientific Chemicals
H5873106
5 g
Each for $437.79
Only null left
 
Description

Description

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
Specifications

Chemical Identifiers

915402-03-8
300.28
LDKJXCOLGXLWKP-UHFFFAOYSA-N
17999151
C17H25BO2Si
MFCD08706285
4,4,5,5-tetramethyl-2-3-trimethylsilanylethynyl-phenyl-1,3,2 dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl-trimethylsilane, trimethyl 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, 3-trimethylsilyl ethynyl phenylboronic acid pinacol ester, trimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, amtb201, 3-2-trimethylsilyl ethynyl benzeneboronic acid pinacol ester
CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C#C[Si](C)(C)C

Specifications

915402-03-8
C17H25BO2Si
1 g
LDKJXCOLGXLWKP-UHFFFAOYSA-N
trimethyl-[2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl]silane
17999151
97%
86°C to 90°C
MFCD08706285
4,4,5,5-tetramethyl-2-3-trimethylsilanylethynyl-phenyl-1,3,2 dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl-trimethylsilane, trimethyl 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, 3-trimethylsilyl ethynyl phenylboronic acid pinacol ester, trimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, amtb201, 3-2-trimethylsilyl ethynyl benzeneboronic acid pinacol ester
CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C#C[Si](C)(C)C
300.28
300.28
3-[2-(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only