missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Thienyl)-L-alanine 97.0+%, TCI America™
$82.02 - $274.02
Chemical Identifiers
| CAS | 22951-96-8 |
|---|---|
| Molecular Formula | C7H9NO2S |
| Molecular Weight (g/mol) | 171.214 |
| MDL Number | MFCD00064335 |
| InChI Key | WTOFYLAWDLQMBZ-LURJTMIESA-N |
| Synonym | 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715 |
| PubChem CID | 146719 |
| IUPAC Name | (2S)-2-amino-3-thiophen-2-ylpropanoic acid |
| SMILES | C1=CSC(=C1)CC(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3475200MG
|
TCI America
T3475200MG |
200 mg |
Each for $82.02
|
|
|||||
|
T34751G
|
TCI America
T34751G |
1 g |
Each for $274.02
|
|
|||||
Chemical Identifiers
| 22951-96-8 | |
| 171.214 | |
| WTOFYLAWDLQMBZ-LURJTMIESA-N | |
| 146719 | |
| C1=CSC(=C1)CC(C(=O)O)N |
| C7H9NO2S | |
| MFCD00064335 | |
| 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715 | |
| (2S)-2-amino-3-thiophen-2-ylpropanoic acid |
Specifications
| 22951-96-8 | |
| C7H9NO2S | |
| 200 mg | |
| WTOFYLAWDLQMBZ-LURJTMIESA-N | |
| (2S)-2-amino-3-thiophen-2-ylpropanoic acid | |
| 146719 | |
| ≥97.0% (HPLC,T) | |
| 3-(2-Thienyl)-L-alanine |
| 235°C | |
| MFCD00064335 | |
| 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715 | |
| C1=CSC(=C1)CC(C(=O)O)N | |
| 171.214 | |
| 171.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No