missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4,4-disulfonic acid sodium salt, ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187869
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015275 1 g
111015276 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015275 Supplier Merck Emd Millipore Supplier No. 829501G
Only null left

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt also known as Ferrozine is a chelating agent for ferrous iron.

Chemical Identifiers

MDL Number MFCD00078417
Synonym 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4 ′,4 ′′-disulfonic acid sodium salt; PDT disulfonate

Specifications

Melting Point ≥300°C
CAS 69898-45-9
Quantity 1 g
Linear Formula C20H13N4 NaO6S2
Formula Weight 492.46
Percent Purity ≥97.0% (T); ≥97.0%