missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5,5-disulfonic acid disodium salt, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111015558
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015558 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015558 Supplier Merck Emd Millipore Supplier No. 829401G

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5',5″-disulfonic acid disodium salt also known as Ferene is a chelating agent which usually binds with Fe2+ ion to form coloured complex.

Chemical Identifiers

MDL Number MFCD00040642
Synonym 5,5 ′-[3-(2-Pyridyl)-1,2,4-triazine-5,6-diyl]difuran-2-sulfonic acid disodium salt

Specifications

CAS 79551-14-7
Linear Formula C16H8N4 Na2O8S2
Quantity 1 g
Formula Weight 494.37
Percent Purity ≥99.0% (TLC); ≥99.0%
Grade For Spectrophotometric Det. of Fe