missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)-5,6-bis(4-sulfophenyl)-1,2,4-triazine Disodium Salt Hydrate 98.0+%, TCI America™

Catalog No. P08301G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P08301G Supplier TCI America Supplier No. P08301G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28048-33-1
Molecular Formula C20H12N4Na2O6S2
Molecular Weight (g/mol) 514.44
MDL Number MFCD00717621
InChI Key DDIHLFCSEHAOJZ-UHFFFAOYSA-L
Synonym ferrozin, ferrozine, ferrozine disodium salt, unii-29j2ciw9pk, 29j2ciw9pk, 3-2-pyridyl-5,6-di 4-phenylsulfonic acid-1,2,4-triazine, disodium salt, 4,4'-3-2-pyridyl-triazine-5,6-diyl dibenzenesulfonic acid disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid, disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid,disodium salt, benzenesulfonic acid, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-, disodium salt
PubChem CID 34127
IUPAC Name disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=CC=N1

Specifications

Quantity 1 g
Formula Weight 514.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Pyridyl)-5,6-bis(4-sulfophenyl)-1,2,4-triazine Disodium Salt Hydrate
RTECSNumber DB7345000
TSCA Yes