missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-D-alanine 98.0+%, TCI America™

Catalog No. p-7133921
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06651G 1 g
N06655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06651G Supplier TCI America Supplier No. N06651G
Only null left

Chemical Identifiers

CAS 76985-09-6
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD00038546
InChI Key JPZXHKDZASGCLU-GFCCVEGCSA-N
Synonym 3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
PubChem CID 2733661
IUPAC Name (2R)-2-azaniumyl-3-(naphthalen-2-yl)propanoate
SMILES [NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 215.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Naphthyl)-D-alanine