missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methylphenyl)propionic acid, 96%

Catalog No. p-7032769
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3437503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3437503 Supplier Thermo Scientific Chemicals Supplier No. H3437503

CAS: 22084-89-5 | C10H12O2 | 164.204 g/mol

3-(2-Methylphenyl)propionic acid is a carboxylic acid building block. It participates in the synthesis of 2-methyl-3-phenylpropanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Methylphenyl)propionic acid is a carboxylic acid building block. It participates in the synthesis of 2-methyl-3-phenylpropanol.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 22084-89-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00079773
InChI Key JIRKNEAMPYVPTD-UHFFFAOYSA-N
PubChem CID 30938
IUPAC Name 3-(2-methylphenyl)propanoic acid
SMILES CC1=CC=CC=C1CCC(=O)O

Specifications

Melting Point 101°C to 103°C
Quantity 1 g
Synonym 3-2-methylphenyl propionic acid, 3-2-methylphenyl propanoic acid, 2-methylhydrocinnamic acid, benzenepropanoic acid, 2-methyl, 3-o-tolyl propanoic acid, 3-o-tolylpropanoic acid, 2-methylbenzenepropanoic acid, benzenepropanoic acid,2-methyl, 2-methylhydrozimtsaure
Solubility Information Soluble in water.
Formula Weight 164.2
Percent Purity 96%
Chemical Name or Material 3-(2-Methylphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only